missing translation for 'onlineSavingsMsg'
Learn More

ML281, MedChemExpress

Product Code. 16451086
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16451086 200 mg 200mg
16438407 5 mg 5mg
16421877 50 mg 50mg
16416206 100 mg 100mg
16405718 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16451086 Supplier Medchem Express Supplier No. HY13495

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ML281 is a potent and selective STK33 inhibitor with IC50 of 14 nM.

ML281 is a potent and selective STK33 inhibitor with IC50 of 14 nM.

Specifications

Chemical Name or Material ML281
CAS 1404437-62-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H19N3O2S
Quantity 200 mg
Solubility Information DMSO : 100 mg/mL (256.76 mM; Need ultrasonic)
SMILES O=C1NC2=CC=CC=C2N=C1C3=CC(C(C)C)=CC=C3NC(C4=CC=CS4)=O
Molecular Weight (g/mol) 389.47
Formula Weight 389.47
Percent Purity 99.88%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.