missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Cynomolgus PVRIG Protein, Fc Tag

Product Code. 16497989
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg

Product Code. 16497989

Brand: ACROBiosystems PVGC52591mg

Please to purchase this item. Need a web account? Register with us today!

Human PVRIG (poliovirus receptor related immunoglobulin domain-containing protein), also known as CD112 receptor (CD112R), is an approximately 34 kDa single transmembrane protein in the poliovirus receptor-like protein (PVR) family. The CD112R gene encodes a putative single transmembrane protein, which is composed of a single extracellular IgV domain, one transmembrane domain, and a long intracellular domain. Notably, the intracellular domain of phatases. The extracellular domain sequence of human and mouse CD112R have 65.3% similarity. CD112R may act as a coinhibitory receptor that suppresses T-cell receptor-mediated signals.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.1 kDa. The protein migrates as 45-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.

Specifications

Accession Number XP_005549281.1
Molecular Weight (g/mol) 40.1 kDa
Quantity 1 mg
Source Cynomolgus PVRIG, Fc Tag (PVG-C5259) is expressed from human 293 cells (HEK293). It contains AA Thr 41 - Asp 171 (Accession # A0A2K5WVV8-1).
Immunogen Thr 41 - Asp 171
Storage Requirements -20°C
Gene Alias C7orf15,C7orf15MGC138295,CD112R,MGC104322,MGC138297,MGC2463,PVRIG,CD112 receptor
Common Name PVRIG
Conjugate Unconjugated
Species Cynomolgus Monkey
Protein Tag Fc Tag
Buffer Tris with Glycine, Arginine and NaCl, pH7.5
Endotoxin Level Less than 1.0 EU / μg by the LAL method.
Expression System HEK293
Form Powder
Protein Subtype Immune Checkpoint Proteins
Purity or Quality Grade >95% as determined by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.