missing translation for 'onlineSavingsMsg'
Learn More

ML204, MedChemExpress

Product Code. p-200096239
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16476036

Brand: Medchem Express HY12949

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ML204 is a potent, selective TRPC4/TRPC5 channel inhibitor, with at least 19-fold selectivity against TRPC6 and no appreciable effect on all other TRP channels, nor on voltage-gated sodium, potassium, or Ca2+ channels.

ML204 is a potent, selective TRPC4/TRPC5 channel inhibitor, with at least 19-fold selectivity against TRPC6 and no appreciable effect on all other TRP channels, nor on voltage-gated sodium, potassium, or Ca2+ channels.

Specifications

Chemical Name or Material ML204
CAS 5465-86-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C15H18N2
Quantity 100 mg
Solubility Information Ethanol : 50 mg/mL (220.93 mM; Need ultrasonic) ∣DMSO : ≥ 37 mg/mL (163.49 mM)
SMILES CC1=CC(N2CCCCC2)=NC3=CC=CC=C13
Molecular Weight (g/mol) 226.32
Formula Weight 226.32
Percent Purity 99.24%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.