missing translation for 'onlineSavingsMsg'
Learn More

ML167, MedChemExpress

Product Code. 16499467
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499467 5 mg 5mg
16446928 10 mg 10mg
16432987 50 mg 50mg
16403516 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16499467 Supplier Medchem Express Supplier No. HY15951
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress ML167 is a highly selective Cdc2-like kinase 4 (Clk4) inhibitor with IC50 of 136 nM, >10-fold selectivity for closely related kinases Clk1, Clk2, Clk3 and Dyrk1A/1B.

ML167 is a highly selective Cdc2-like kinase 4 (Clk4) inhibitor with IC50 of 136 nM, >10-fold selectivity for closely related kinases Clk1, Clk2, Clk3 and Dyrk1A/1B.

Specifications

Chemical Name or Material ML167
CAS 1285702-20-6
Color Khaki
Purity Grade Notes Research
Molecular Formula C19H17N3O3
Quantity 5 mg
Synonym CID44968231 NCGC00188654
Solubility Information DMSO : 16.67 mg/mL (49.71 mM; Need ultrasonic)
SMILES OCC1=CC=C(C2=CC3=C(NCC4=CC=C(C)O4)N=CN=C3C=C2)O1
Molecular Weight (g/mol) 335.36
Formula Weight 335.36
Percent Purity 98.08%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.