missing translation for 'onlineSavingsMsg'
Learn More

ML133 hydrochloride, MedChemExpress

Product Code. 16492858
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16492858 10 mg 10mg
16474266 100 mg 100mg
16439516 50 mg 50mg
16415607 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16492858 Supplier Medchem Express Supplier No. HY100230A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ML133 hydrochloride is a selective Kir2 family channels inhibitor, with an IC50 of 1.8 μM at pH 7.4 and 290 nM at pH 8.5.

ML133 hydrochloride is a selective Kir2 family channels inhibitor, with an IC50 of 1.8 μM at pH 7.4 and 290 nM at pH 8.5.

Specifications

Chemical Name or Material ML133 hydrochloride
CAS 1222781-70-5
Color White
Purity Grade Notes Research
Molecular Formula C19H20ClNO
Quantity 10 mg
Solubility Information DMSO : ≥ 125 mg/mL (398.32 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES COC1=CC=C(CNCC2=C3C=CC=CC3=CC=C2)C=C1.[H]Cl
Molecular Weight (g/mol) 313.82
Formula Weight 313.82
Percent Purity 99.9%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.