missing translation for 'onlineSavingsMsg'
Learn More

ML133 hydrochloride, MedChemExpress

Product Code. 16439516
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16439516

Brand: Medchem Express HY100230A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ML133 hydrochloride is a selective Kir2 family channels inhibitor, with an IC50 of 1.8 μM at pH 7.4 and 290 nM at pH 8.5.

ML133 hydrochloride is a selective Kir2 family channels inhibitor, with an IC50 of 1.8 μM at pH 7.4 and 290 nM at pH 8.5.

Specifications

Chemical Name or Material ML133 hydrochloride
CAS 1222781-70-5
Color White
Purity Grade Notes Research
Molecular Formula C19H20ClNO
Quantity 50 mg
Solubility Information DMSO : ≥ 125 mg/mL (398.32 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES COC1=CC=C(CNCC2=C3C=CC=CC3=CC=C2)C=C1.[H]Cl
Molecular Weight (g/mol) 313.82
Formula Weight 313.82
Percent Purity 99.9%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.