missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ML T7
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
ML T7 is a T cell immunoglobulin and mucin-containing molecule 3 (Tim-3) inhibitor; disrupts phosphatidylserine/CAECAM1 binding to Tim-3 (Kd = 7.0 μM and 7.4 μM for hTim-3 and mTim-3 respectively). ML T7 promotes cell proliferation, cytokine production and cytotoxicity of OT-I CD8+ cytotoxic T lymphocytes. ML T7 inhibits tumor growth in a syngeneic mouse model of hepatocellular carcinoma, increases NK cell tumor killing activity and DC maturation.
Specifications
Specifications
| Chemical Name or Material | 5-(2,4-Dichlorophenyl)-3a,6a-dihydro-3-(4-methylphenyl)spiro[1H-furo[3,4-c]pyrrole-1,2'-[2H]indene]-1',3',4,6(3H,5H)-tetrone |
| CAS | 459789-75-4 |
| Quantity | 50 mg |
| Target | CEACAM1/CD66a |
| Molecular Formula | C27H17Cl2NO5 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction