missing translation for 'onlineSavingsMsg'
Learn More

ML SA1, TRC

Product Code. 30479050
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30479050 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30479050 Supplier Toronto Research Chemicals Supplier No. TRCM42547050MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[2-(3,4-Dihydro-2,2,4-trimethyl-1(2H)-quinolinyl)-2-oxoethyl]-1H-isoindole-1,3(2H)-dione
CAS 332382-54-4
Molecular Formula C22H22N2O3
Quantity 50 mg
Synonym ML SA1, 2-[2-(3,4-Dihydro-2,2,4-trimethyl-1(2H)-quinolinyl)-2-oxoethyl]-1H-isoindole-1,3(2H)-dione
InChI Formula InChI=1S/C22H22N2O3/c1-14-12-22(2,3)24(18-11-7-6-8-15(14)18)19(25)13-23-20(26)16-9-4-5-10-17(16)21(23)27/h4-11,14H,12-13H2,1-3H3
SMILES CC1CC(C)(C)N(C(=O)CN2C(=O)c3ccccc3C2=O)c4ccccc14
IUPAC Name 2-[2-oxo-2-(2,2,4-trimethyl-3,4-dihydroquinolin-1-yl)ethyl]isoindole-1,3-dione
Molecular Weight (g/mol) 362.42
Formula Weight 362.163
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.