missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ML 9 hydrochloride
GREENER_CHOICE

Product Code. 30000900
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30000900 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30000900 Supplier Tocris Bioscience™ Supplier No. 0431/10
  • ONLINE EXCLUSIVE PRICE
    £161.00
    £152.25 / 10mg
    Save £8.75
    5% Off
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Myosin light chain kinase inhibitor

ML 9 hydrochloride is a selective MLCK inhibitor (Ki values are 4, 32 and 54 μM for MLCK, PKA and PKC respectively). Also inhibits STIM1-plasma membrane interactions, preventing store-operated Ca2+ entry (SOCE).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105637-50-1
Molecular Formula C15H18Cl2N2O2S

Specifications

Quantity 10 mg
Formula Weight 361.29
Purity >99%
Molecular Weight (g/mol) 361.281
Chemical Name or Material ML 9 hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.