missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ML 7 hydrochloride

Description
Selective MLCK inhibitor (Ki = 0.3 μM). Exhibits more potent inhibition than ML 9 hydrochloride (Cat. No. 0431) Displays reversible, ATP-competitive inhibition of Ca2+-calmodulin-dependent and -independent smooth muscle MLCKs. Inhibits proplatelet formation and stabilization.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 452.74 |
| Purity | 0.98 |
| Molecular Weight (g/mol) | 452.735 |
| Chemical Name or Material | ML 7 hydrochloride |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?