missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ML 7 hydrochloride
GREENER_CHOICE

Product Code. 18015592
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18015592 10 mg 10mg
18034412 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18015592 Supplier Tocris Bioscience™ Supplier No. 4310/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective inhibitor of myosin light chain kinase

Selective MLCK inhibitor (Ki = 0.3 μM). Exhibits more potent inhibition than ML 9 hydrochloride (Cat. No. 0431) Displays reversible, ATP-competitive inhibition of Ca2+-calmodulin-dependent and -independent smooth muscle MLCKs. Inhibits proplatelet formation and stabilization.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110448-33-4
Molecular Formula C15H18ClIN2O2S

Specifications

Quantity 10 mg
Formula Weight 452.74
Purity 0.98
Molecular Weight (g/mol) 452.735
Chemical Name or Material ML 7 hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.