missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ML 418
GREENER_CHOICE

Product Code. 18715946
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18715946

Brand: Tocris Bioscience™ 6889/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Kir 7.1 inhibitor. Also inhibits Kir 6.2

ML 418 is a Kir7.1 inhibitor (IC50 = 0.31 μM). Also inhibits Kir6.2 with similar potency. Displays >17-fold selectivity for Kir7.1 over Kir1.1, Kir2.1, Kir2.2, Kir2.3, Kir3.1/3.2 and Kir4.1. Active in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isopropyl (1-((5-chloro-8-hydroxyquinolin-7-yl)methyl)piperidin-4-yl)carbamate
CAS 1928763-08-9
Quantity 10 mg
Molecular Formula C19H24CIN3O3
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.