missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ML 348
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
ML 348 is a selective and reversible lysophospholipase 1 (LYPLA1) inhibitor (IC50 = 210 nM). Exhibits 14-fold selectivity for LYPLA1 over LYPLA2. Also selective over a panel of ∽30 other serine hydrolases.
Specifications
Specifications
| Purity | >99% |
| Quantity | 10 mg |
| Product Type | ML 348 |
| Molecular Weight (g/mol) | 415.79 |
| Inhibitors | ML 348 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction