missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ML 347

Product Code. 18725718
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18725718 50 mg 50mg
18788617 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18725718 Supplier Tocris Bioscience™ Supplier No. 4945/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective ALK1 and ALK2 inhibitor

ML 347 is a potent and selective ALK2 and ALK1 inhibitor (IC50 values are 32 and 46 nM, respectively); displays >200 fold selectivity over ALK3 and ALK6 and >400 fold selectivity over VEGF2. Exhibits no activity at ALK4, ALK5 or in a panel of related kinases. Inhibits BMP4 signaling in a functional assay (IC50 = 152 nM).
TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 mg
Product Type ML 347
Purity >98%
Molecular Weight (g/mol) 352.39
Inhibitors ML 347

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.