missing translation for 'onlineSavingsMsg'
Learn More

ML-323, MedChemExpress

Product Code. 16499857
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499857 5 mg 5mg
16487388 10 mg 10mg
16483417 50 mg 50mg
16407366 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16499857 Supplier Medchem Express Supplier No. HY17543
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress ML-323 is a reversible, potent USP1-UAF1 inhibitor with IC50 of 76 nM in a Ub-Rho assay. The measured inhibition constants of ML-323 for the free enzyme (Ki) is 68 nM.

ML-323 is a reversible, potent USP1-UAF1 inhibitor with IC50 of 76 nM in a Ub-Rho assay. The measured inhibition constants of ML-323 for the free enzyme (Ki) is 68 nM.

Specifications

Chemical Name or Material ML-323
CAS 1572414-83-5
Color White
Purity Grade Notes Research
Molecular Formula C23H24N6
Quantity 5 mg
Solubility Information DMSO : ≥ 49 mg/mL (127.44 mM)
SMILES CC1=CN=C(C2=C(C(C)C)C=CC=C2)N=C1NCC3=CC=C(N4N=NC=C4)C=C3
Molecular Weight (g/mol) 384.48
Formula Weight 384.48
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.