missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ML 315 hydrochloride
GREENER_CHOICE

Product Code. 18774539
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18774539

Brand: Tocris Bioscience™ 5700/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Low molecular weight, organic compound that may help regulate biological process.

ML 315 hydrochloride is a inhibitor of cdc2-like kinases (Clk) and DYRK kinases (IC50 values are 68, 68 and 231 nM for Clk1, Clk4 and Clk2, respectively, and 282 and 1156 nM for Dyrk1A and Dyrk1B). Shows 20-fold selectivity for Clk4 over Dyrk1A. Stable in both mouse and human plasma. Moderately cell permeable.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1440251-53-5
Quantity 10 mg
Target Cdc2-like Kinase
Molecular Formula C18H13Cl2N3O2.HCl
Formula Weight 410.68g/mol
Product Type Inhibitor

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.