missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ML 311
Description
ML 311 is an inhibits Mcl1-BIM interaction. Inhibits viability of Mcl1-dependent tumor cell lines. Induces cell death and decreased spheroid formation in Her2 positive breast cancer cells.
Specifications
Specifications
| Chemical Name or Material | 7-[(4-Ethyl-1-piperazinyl)[4-(trifluoromethyl)phenyl]methyl]-8-quinolinol |
| CAS | 315698-17-0 |
| Assay Percent Range | >99% |
| Quantity | 50 mg |
| Molecular Formula | C23H24F3N3O |
| Formula Weight | Observed MW: 415.45 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?