missing translation for 'onlineSavingsMsg'
Learn More

ML 311

Product Code. 18748078
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18748078 10 mg 10mg
18796178 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18748078 Supplier Tocris Bioscience Supplier No. 5733/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Inhibits Mcl-1-BIM interaction

ML 311 is an inhibits Mcl1-BIM interaction. Inhibits viability of Mcl1-dependent tumor cell lines. Induces cell death and decreased spheroid formation in Her2 positive breast cancer cells.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 7-[(4-Ethyl-1-piperazinyl)[4-(trifluoromethyl)phenyl]methyl]-8-quinolinol
CAS 315698-17-0
Assay Percent Range >99%
Quantity 10 mg
Molecular Formula C23H24F3N3O
Formula Weight Observed MW: 415.45
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.