missing translation for 'onlineSavingsMsg'
Learn More

ML-290, MedChemExpress

Product Code. p-200101360
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16444658

Brand: Medchem Express HY112606

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ML-290 is a first-in-class and potent relaxin/insulin-like family peptide receptor (RXFP1) agonist and activator of anti-fibrotic genes, with an EC50 of 94 nM. ML290 is a biased allosteric agonist at the relaxin receptor RXFP1.

ML-290 is a first-in-class and potent relaxin/insulin-like family peptide receptor (RXFP1) agonist and activator of anti-fibrotic genes, with an EC50 of 94 nM. ML290 is a biased allosteric agonist at the relaxin receptor RXFP1.

Specifications

Chemical Name or Material ML-290
CAS 1482500-76-4
Color White
Purity Grade Notes Research
Molecular Formula C24H21F3N2O5S
Quantity 10 mg
Solubility Information DMSO : 125 mg/mL (246.80 mM; Need ultrasonic)
SMILES O=C(NC1=CC(S(=O)(C(F)(F)F)=O)=CC=C1)C2=C(NC(C3=CC=CC=C3OC(C)C)=O)C=CC=C2
Molecular Weight (g/mol) 506.49
Formula Weight 506.49
Percent Purity 98.0%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.