missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ML 221
GREENER_CHOICE

Product Code. 30001685
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001685 50 mg 50mg
30001666 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001685 Supplier Tocris Bioscience™ Supplier No. 4748/50
  • ONLINE EXCLUSIVE PRICE
    £879.00
    £830.55 / 50mg
    Save £48.45
    5% Off
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Apelin antagonist

Apelin receptor (APJ) antagonist (IC50 values are 0.70 and 1.75 μM in a cAMP assay and β-arrestin assay, respectively). Displays >37-fold selectivity over the closely related angiotensin II type 1 (AT1) receptor. Exhibits no toxicity towards human hepatocytes at concentrations >50 μM.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 877636-42-5
Molecular Formula C17H11N3O6S

Specifications

Quantity 50 mg
Formula Weight 385.35
Purity >99%
Molecular Weight (g/mol) 385.35
Chemical Name or Material ML 221
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.