missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ML 184
GREENER_CHOICE

Product Code. 18788536
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18788536

Brand: Tocris Bioscience™ 6668/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective GPR55 agonist; also promotes NSC proliferation and differentiation

ML 184 is a selective GPR55 agonist (EC50 = 250 nM). Exhibits >100-fold selectivity for GPR55 over GPR35, CB1 and CB2. Increases proliferation of neural stem cells and promotes neuronal differentiation in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Synonym CID 20440433
Quantity 10 mg
Molecular Formula C25H34N4O3S
Formula Weight 470.63
Product Type Agonist
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.