missing translation for 'onlineSavingsMsg'
Learn More

ML-030, TRC

Product Code. 30477431
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30477431 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30477431 Supplier Toronto Research Chemicals Supplier No. TRCM42505510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(2,5-dimethoxyphenyl)-6-(3,4-dimethoxyphenyl)-7H-1,2,4-triazolo[3,4-b][1,3,4]thiadiazine
CAS 1013750-77-0
Molecular Formula C20H20N4O4S
Quantity 10 mg
Synonym CID-11757146, 3-(2,5-dimethoxyphenyl)-6-(3,4-dimethoxyphenyl)-7H-1,2,4-triazolo[3,4-b][1,3,4]thiadiazine
InChI Formula InChI=1S/C20H20N4O4S/c1-25-13-6-8-16(26-2)14(10-13)19-21-22-20-24(19)23-15(11-29-20)12-5-7-17(27-3)18(9-12)28-4/h5-10H,11H2,1-4H3
SMILES COC1=CC(C2=NN=C3SCC(C4=CC(OC)=C(OC)C=C4)=NN23)=C(OC)C=C1
IUPAC Name 3-(2,5-dimethoxyphenyl)-6-(3,4-dimethoxyphenyl)-7H-[1,2,4]triazolo[3,4-b][1,3,4]thiadiazine
Molecular Weight (g/mol) 412.46
Formula Weight 412.12
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.