missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MKS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576177
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The protein encoded by this gene localizes to the basal body and is required for formation of the primary cilium in ciliated epithelial cells. Mutations in this gene result in Meckel syndrome type 1 and in Bardet-Biedl syndrome type 13. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| MKS1 | |
| Polyclonal | |
| Unconjugated | |
| MKS1 | |
| AK190930; B8d3; BBS13; LOW QUALITY PROTEIN: Meckel syndrome type 1 protein; Meckel syndrome type 1 protein; Meckel syndrome type 1 protein homolog; Meckel syndrome, type 1; MES; MKS; MKS transition zone complex subunit 1; Mks1; POC12; POC12 centriolar protein homolog | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 287612, 380718, 54903 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q499Q5, Q5SW45, Q9NXB0 | |
| MKS1 | |
| Recombinant protein of human MKS1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction