missing translation for 'onlineSavingsMsg'
Learn More

MKRN3 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18404255 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18404255 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18404255 Supplier Novus Biologicals Supplier No. NBP281964AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MKRN3 Polyclonal antibody specifically detects MKRN3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen MKRN3
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias EC 6.3.2.-, makorin ring finger protein 3, MGC88288, RNF63D15S9, ZFP127RING finger protein 63, Zinc finger protein 127, ZNF127probable E3 ubiquitin-protein ligase makorin-3
Host Species Rabbit
Immunogen MKRN3 antibody was raised against a 17 amino acid peptide near the carboxy terminus of human MKRN3. The immunogen is located within amino acids 400 - 450 of MKRN3. Amino Acid Squence: FYKHEYPEGWGDEPPG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 7681
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.