missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MKRN2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-17302
This item is not returnable.
View return policy
Description
MKRN2 Polyclonal specifically detects MKRN2 in Human samples. It is validated for Western Blot.
Specifications
| MKRN2 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| EC 6.3.2.-, HSPC070, makorin ring finger protein 2, probable E3 ubiquitin-protein ligase makorin-2, RNF62RING finger protein 62 | |
| Rabbit | |
| 47 kDa | |
| 0.1 mL | |
| Signal Transduction, Zinc Finger | |
| 23609 | |
| Human | |
| IgG |
| Western Blot | |
| 0.74mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q9H000 | |
| MKRN2 | |
| Recombinant protein encompassing a sequence within the center region of human MKRN2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Species reactivity based on sequence identity: Mouse (82%), Bovine (84%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction