missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MKP4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582081
This item is not returnable.
View return policy
Description
Immunogen sequence: ECPHLCETSL AGRAGSSMAP VPGPVPVVGL GSLCLGSDCS DAESEADRDS MSCGLDSEGA TPPPVGLRAS FPVQILPNLY LGSARDSANL ESLAKLGIRY ILNVTPNLPN FFEKNGDFHY KQI Highest antigen sequence indentity to the following orthologs: Mouse - 82%, Rat - 80%.
Mitogen-activated protein (MAP) kinases are a large class of proteins involved in signal transduction pathways that are activated by a range of stimuli and mediate a number of physiological and pathological changes in the cell. Dual specificity phosphatases (DSPs) are a subclass of the protein tyrosine phosphatase (PTP) gene superfamily, which are selective for dephosphorylating critical phosphothreonine and phosphotyrosine residues within MAP kinases. DSP gene expression is induced by a host of growth factors and/or cellular stresses, thereby negatively regulating MAP kinase superfamily members including MAPK/ERK, SAPK/JNK and p38. The members of the dual-specificity phosphatase protein family include MKP-1/CL100 (3CH134), VHR, PAC1, MKP-2, hVH-3 (B23), hVH-5, MKP-3, MKP-X and MKP-4.
Specifications
| MKP4 | |
| Polyclonal | |
| Unconjugated | |
| DUSP9 | |
| dual specificity phosphatase 9; dual specificity protein phosphatase 9; dual-specificity MAP kinase phosphatase 4; Dusp4; DUSP9; MAP kinase phosphatase 4; mitogen-activated protein kinase phosphatase 4; MKP4; MKP-4; Mpk4; Pyst3; serine/threonine specific protein phosphatase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1852 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q99956 | |
| DUSP9 | |
| Recombinant protein corresponding to Human DUSP9. Recombinant protein control fragment (Product #RP-92206). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction