missing translation for 'onlineSavingsMsg'
Learn More

MKP-3/DUSP6 Antibody (363209) [Janelia Fluor™ 669], Novus Biologicals™

Product Code. 30736001 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30736001 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736001 Supplier Novus Biologicals Supplier No. FAB35761JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MKP-3/DUSP6 Monoclonal antibody specifically detects MKP-3/DUSP6 in Human samples. It is validated for Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen MKP-3/DUSP6
Applications Immunocytochemistry
Classification Monoclonal
Clone 363209
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias dual specificity phosphatase 6, Dual specificity protein phosphatase PYST1, EC 3.1.3.16, EC 3.1.3.48, MAP kinase phosphatase 3, Mitogen-activated protein kinase phosphatase 3, MKP-3, MKP3serine/threonine specific protein phosphatase, PYST1dual specificity protein phosphatase 6
Host Species Mouse
Immunogen E. coli-derived recombinant human MKP-3, Met1-Thr381, Accession # Q16828
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 1848
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.