missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MKL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588993
This item is not returnable.
View return policy
Description
Immunogen sequence: IIGVGKEDYP HTQGDFSFDE DSSDALSPDQ PASQESQGSA ASPSEPKVSE SPSPVTTNTP AQFASVSPTV PEFLKTPPTA DQPPPRPAAP VLPTNTVSSA KPGPALVKQS HPKNPNDKHR SKKCKDPKPR V; Positive Samples: LO2, Jurkat, Mouse pancreas; Cellular Location: Nucleus.
Serum response factor (SRF) is a transcription factor that binds the serum response element (SRE), a sequence that mediates the transient response of many cellular genes to growth stimulation. SRF regulates the transient response of several muscle genes in response to growth factors and recruits accessory myogenic factors to activate these muscle genes. SRF is required for the formation of vertebrate mesoderm leading to the origin of the cardiovascular system. Myocardin, in association with SRF in cardiac muscle cells, activates cardiac muscle promoters. Myocardin-related transcription factors A and B (MRTF-A and MRTF-B) interact with SRF and act as stimulators for its transcriptional activity. MRTF-B is crucial for skeletal myogenic differentiation.
Specifications
| MKL2 | |
| Polyclonal | |
| Unconjugated | |
| Mkl2 | |
| CTA-276F8.1; ENSMUSG00000075401; gene trap insertion site 4-1; Gt4-1; KIAA1243; Megakaryoblastic leukemia 2; megakaryoblastic leukemia 2 protein; mKIAA1243; MKL/myocardin like 2; MKL/myocardin-like 2; MKL/myocardin-like protein 2; MKL1/myocardin like 2; Mkl2; Mrtfb; MRTF-B; myocardin related transcription factor B; myocardin-related transcription factor B; myocardin-related transcription factor B; MKL/myocardin-like protein 2; NPD001; RGD1560833 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 239719, 57496 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.07 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P59759, Q9ULH7 | |
| Mkl2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 170-300 of human MKL2 (NP_0547673). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction