missing translation for 'onlineSavingsMsg'
Learn More

MKL2 Antibody, DyLight 755, Novus Biologicals™

Product Code. 18433295 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18433295 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18433295 Supplier Novus Biologicals Supplier No. NBP146208IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MKL2 Polyclonal antibody specifically detects MKL2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MKL2
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 755
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias DKFZp686J1745, FLJ31823, FLJ45623, KIAA1243, megakaryoblastic leukemia 2, MKL/myocardin-like 2, MKL/myocardin-like protein 2, MRTFB, MRTF-B, myocardin-related transcription factor B, NPD001
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 900 and 950 of human Megakaryoblastic Leukemia 2 using the numbering given in entry NP_054767.3 (GeneID 57496).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 57496
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.