missing translation for 'onlineSavingsMsg'
Learn More

MKL2 Antibody (CL1546), Novus Biologicals™

Product Code. 18697528 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18697528 25 μL 25µL
18606947 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18697528 Supplier Novus Biologicals Supplier No. NBP25297025ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MKL2 Monoclonal antibody specifically detects MKL2 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen MKL2
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone CL1546
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL
Formulation PBS (pH 7.2), 40% Glycerol
Gene Alias DKFZp686J1745, FLJ31823, FLJ45623, KIAA1243, megakaryoblastic leukemia 2, MKL/myocardin-like 2, MKL/myocardin-like protein 2, MRTFB, MRTF-B, myocardin-related transcription factor B, NPD001
Host Species Mouse
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: QRPGPMELVEKNILPVDSSVKEAIIGVGKEDYPHTQGDFSFDEDSSDALSPDQPASQESQGSAASPSEPKVSESPSPVTTNTPAQFASVSPTVPEFLKTPPTADQPPPRPAAPVLPTNTVSSAKPGPALVKQSHPKNP
Purification Method Protein A purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 57496
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.