missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MKL2 Antibody (CL1546), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52970
This item is not returnable.
View return policy
Description
MKL2 Monoclonal antibody specifically detects MKL2 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| MKL2 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence | |
| CL1546 | |
| Western Blot 1 μg/mL, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL | |
| DKFZp686J1745, FLJ31823, FLJ45623, KIAA1243, megakaryoblastic leukemia 2, MKL/myocardin-like 2, MKL/myocardin-like protein 2, MRTFB, MRTF-B, myocardin-related transcription factor B, NPD001 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: QRPGPMELVEKNILPVDSSVKEAIIGVGKEDYPHTQGDFSFDEDSSDALSPDQPASQESQGSAASPSEPKVSESPSPVTTNTPAQFASVSPTVPEFLKTPPTADQPPPRPAAPVLPTNTVSSAKPGPALVKQSHPKNP | |
| 0.1 mL | |
| Cardiovascular Biology, Stem Cells | |
| 57496 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction