Learn More
MKL2 Antibody (CL1546) [Alexa Fluor™ 532], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52970AF532
Product Code. 30295940
  • £558.00 / 0.10mL
Description
MKL2 Monoclonal antibody specifically detects MKL2 in Human samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MKL2
Monoclonal
Alexa Fluor 532
DKFZp686J1745, FLJ31823, FLJ45623, KIAA1243, megakaryoblastic leukemia 2, MKL/myocardin-like 2, MKL/myocardin-like protein 2, MRTFB, MRTF-B, myocardin-related transcription factor B, NPD001
This antibody was developed using a recombinant protein derived from Q9ULH7, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cardiovascular Biology, Stem Cells
57496
Store at 4⪚C in the dark.
IgG1
Western Blot, Immunocytochemistry/Immunofluorescence
CL1546
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified