missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MKL1, Mouse anti-Human, Clone: CL4281, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB15707.100uL
This item is not returnable.
View return policy
Description
Sequence: EQEKRAQQPAPAPAPLGTPVKQENSFSSCQLSQQPLGPAHPFNPSLAAPATNHIDPCAVAPGPPSVVVKQEALQPEPEPVPAPQLLLGPQGPSLIKGVAPPTLITDSTGTHLVLTVTNKNADSPGSpecifications
| MKL1 | |
| Monoclonal | |
| Unconjugated | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide). | |
| BSAC/MAL/MRTF-A | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| CL4281 | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-1:200) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| megakaryoblastic leukemia (translocation) 1 | |
| MKL1 | |
| Recombinant protein corresponding to human MKL1. | |
| RUO | |
| 57591 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction