missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MKKS Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA560653

Product Code. 15718481

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: KSTALKVALF CTTLSGDTSD TGEGTVVVSY GVSLENAVLD QLLNLGRQLI SDHVDLVLCQ KVIHPSLKQF LNMHRIIAID RIGVTLMEPL TK Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 80%.

Probable molecular chaperone. Assists the folding of proteins upon ATP hydrolysis. As part of the BBS/CCT complex may play a role in the assembly of BBSome, a complex involved in ciliogenesis regulating transports vesicles to the cilia. May play a role in protein processing in limb, cardiac and reproductive system development. May play a role in cytokinesis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MKKS
Polyclonal
Unconjugated
MKKS
1300013E18Rik; AI463362; AI957237; Bardet-Biedl syndrome 6 protein; BBS6; HMCS; KMS; McKusick-Kaufman syndrome; McKusick-Kaufman syndrome protein; McKusick-Kaufman syndrome protein; MKKS protein; McKusick-Kaufman/Bardet-Biedl syndromes putative chaperonin; MKKS; MKS; protein Bbs6 homolog
Rabbit
Antigen affinity chromatography
RUO
8195
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9NPJ1
MKKS
Recombinant protein corresponding to Human MKKS. Recombinant protein control fragment (Product #RP-99409).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.