missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MKKS Polyclonal Antibody

MKKS Polyclonal Antibody
Brand: Invitrogen PA550453
This item is not returnable.
View return policy
Description
MKKS Polyclonal Antibody for IHC (P)
Specifications
| MKKS | |
| Polyclonal | |
| Unconjugated | |
| MKKS | |
| 1300013E18Rik; AI463362; AI957237; Bardet-Biedl syndrome 6 protein; BBS6; HMCS; KMS; McKusick-Kaufman syndrome; McKusick-Kaufman syndrome protein; McKusick-Kaufman syndrome protein; MKKS protein; McKusick-Kaufman/Bardet-Biedl syndromes putative chaperonin; MKKS; MKS; protein Bbs6 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8195 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 2.1 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9NPJ1 | |
| MKKS | |
| Fusion protein corresponding to residues near the C terminal of human McKusick-Kaufman syndrome. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction