missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX MKK6/SKK3 (S599D, T603D) Protein, Active

Product Code. 17360314
Encompass_Preferred
Click to view available options
Quantity:
10 μg
250 μg
Unit Size:
10µg
250µg
This item is not returnable. View return policy

Product Code. 17360314

Brand: Eurofins DiscoverX 14537

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Active, N-terminal MBP tagged, recombinant, human MKK6/SKK3 residues 4-end containing S599D and T603D mutations, for use in Kinase Assays.

MKK6/SKK3 Protein is a member of the dual specificity protein kinase family, which functions as a mitogen-activated protein (MAP) kinase kinase. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals. This protein phosphorylates and activates p38 MAP kinase in response to inflammatory cytokines or environmental stress. As an essential component of p38 MAP kinase mediated signal transduction pathway, this gene is involved in many cellular processes such as stress induced cell cycle arrest, transcription activation and apoptosis.

EMBL U39657

Specifications

Formulation mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol, 270 mM sucrose. Frozen solution.
Molecular Weight (g/mol) 80.6 kDa
Quantity 10 μg
Species Human
Recombinant Recombinant
Protein Tag N-terminal maltose-binding protein (MBP)
Expression System Recombinant enzyme expressed in E. coli cells
Protein Form Truncated
Purity or Quality Grade ≥61%
Protein MKK6/SKK3 (S599D,T603D), active
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.