Learn More
Eurofins DiscoverX MKK6/SKK3 Protein, Active
Description
MKK6/SKK3 Protein is a member of the dual specificity protein kinase family, which functions as a mitogen-activated protein (MAP) kinase kinase. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals. This protein phosphorylates and activates p38 MAP kinase in response to inflammatory cytokines or environmental stress. As an essential component of p38 MAP kinase mediated signal transduction pathway, this gene is involved in many cellular processes such as stress induced cell cycle arrest, transcription activation and apoptosis.
EMBL U39657
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol, 270 mM sucrose. Frozen solution. |
| Molecular Weight (g/mol) | 82 kDa |
| Quantity | 10 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal maltose-binding protein domain |
| Expression System | Recombinant enzyme expressed in E. coli cells |
| Protein Form | Truncated |
| Purity or Quality Grade | ≥73% |
| Protein | MKK6/SKK3 (4-end), active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.