missing translation for 'onlineSavingsMsg'
Learn More

MKK4/MEK4 Antibody (OTI8A8), mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18414148 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18414148 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18414148 Supplier Novus Biologicals Supplier No. NBP272710MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MKK4/MEK4 Monoclonal antibody specifically detects MKK4/MEK4 in Human, Mouse, Rat, Canine samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen MKK4/MEK4
Applications Flow Cytometry
Classification Monoclonal
Clone OTI8A8
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias c-Jun N-terminal kinase kinase 1, dual specificity mitogen-activated protein kinase kinase 4, JNK activating kinase 1, JNK-activated kinase 1, JNK-activating kinase 1, JNKK, JNKK1MAPK/ERK kinase 4, MAP kinase kinase 4, MAPKK4, MEK4MEK 4, mitogen-activated protein kinase kinase 4, MKK4SAPK/ERK kinase 1, PRKMK4MAPKK 4, SEK1, SERK1EC 2.7.12.2
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human MAP2K4 expression vector
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, MAP Kinase Signaling, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 6416
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.