missing translation for 'onlineSavingsMsg'
Learn More

MKK4/MEK4 Antibody (323042) [DyLight 488], Novus Biologicals™

Product Code. 30732174 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30732174 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732174 Supplier Novus Biologicals Supplier No. FAB3390K

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MKK4/MEK4 Monoclonal antibody specifically detects MKK4/MEK4 in Human samples. It is validated for Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen MKK4/MEK4
Applications Immunocytochemistry
Classification Monoclonal
Clone 323042
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias c-Jun N-terminal kinase kinase 1, dual specificity mitogen-activated protein kinase kinase 4, JNK activating kinase 1, JNK-activated kinase 1, JNK-activating kinase 1, JNKK, JNKK1MAPK/ERK kinase 4, MAP kinase kinase 4, MAPKK4, MEK4MEK 4, mitogen-activated protein kinase kinase 4, MKK4SAPK/ERK kinase 1, PRKMK4MAPKK 4, SEK1, SERK1EC 2.7.12.2
Host Species Mouse
Immunogen E. coli-derived recombinant human MKK4, Met1-Asp399, Accession # P45985
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, MAP Kinase Signaling, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 6416
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.