missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MKK4/MEK4 Antibody (2 E4), Novus Biologicals™
Mouse Monoclonal Antibody
£149.00
Specifications
| Antigen | MKK4/MEK4 |
|---|---|
| Clone | 2 E4 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:50 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Description
MKK4/MEK4 Monoclonal specifically detects MKK4/MEK4 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| MKK4/MEK4 | |
| 1 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Cancer, MAP Kinase Signaling, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| c-Jun N-terminal kinase kinase 1, dual specificity mitogen-activated protein kinase kinase 4, JNK activating kinase 1, JNK-activated kinase 1, JNK-activating kinase 1, JNKK, JNKK1MAPK/ERK kinase 4, MAP kinase kinase 4, MAPKK4, MEK4MEK 4, mitogen-activated protein kinase kinase 4, MKK4SAPK/ERK kinase 1, PRKMK4MAPKK 4, SEK1, SERK1EC 2.7.12.2 | |
| MAP2K4 | |
| IgG | |
| Protein A purified |
| 2 E4 | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 6416 | |
| Protein expressed in 293T cells transfected with human MAP2K4 expression vector | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title