missing translation for 'onlineSavingsMsg'
Learn More

MKC3946, MedChemExpress

Product Code. 16470318
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16470318 5 mg 5mg
16451849 1 mg 1mg
16437858 10 mg 10mg
16417676 100 mg 100mg
16413807 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16470318 Supplier Medchem Express Supplier No. HY19710
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress MKC3946 is a potent IRE1α inhibitor, used for cancer research.

MKC3946 is a potent IRE1α inhibitor, used for cancer research.

Specifications

Chemical Name or Material MKC3946
CAS 1093119-54-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H20N2O3S
Quantity 5 mg
Solubility Information DMSO : 20 mg/mL (52.57 mM; Need ultrasonic)
SMILES O=CC1=C(O)C=CC2=C1C=CC(C3=CC=C(C(N4CCN(C)CC4)=O)S3)=C2
Molecular Weight (g/mol) 380.46
Formula Weight 380.46
Percent Purity 99.64%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.