missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ MK 886
Description
MK 886 is a potent inhibitor of 5-lipoxygenase-activating protein (FLAP) (IC50 = 30 nM for inhibition of [125I]-L-691,678 photoaffinity labelling). Inhibits leukotriene biosynthesis (IC50 = 3 nM in human polymorphonuclear leukocytes). Also moderately potent PPARα antagonist (IC50 = 0.5-1 μM). Orally active in vivo.
Specifications
Specifications
| Chemical Name or Material | 1-[(4-Chlorophenyl)methyl]-3-[(1,1-dimethylethyl)thio]-α,α-dimethyl-5-(1-methylethyl)-1H-Indole-2-propanoic acid |
| CAS | 118414-82-7 |
| Quantity | 2 mg |
| Target | Lipoxygenase Inhibitors |
| Molecular Formula | C27H34ClNO2S |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?