missing translation for 'onlineSavingsMsg'
Learn More

MK-886, MedChemExpress

Product Code. 16453406
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16453406 25 mg 25mg
16445978 10 mg 10mg
16438597 5 mg 5mg
16426406 100 mg 100mg
16402127 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16453406 Supplier Medchem Express Supplier No. HY14166
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress MK-886 (L 663536) is a potent, cell-permeable and orally active FLAP (IC50 of 30 nM) and leukotriene biosynthesis (IC50s of 3 nM and 1.1 μM in intact leukocytes and human whole blood, respectively) inhibitor. MK-886 is also a non-competitive PPARα antagonist and can induce apoptosis.

MK-886 (L 663536) is a potent, cell-permeable and orally active FLAP (IC50 of 30 nM) and leukotriene biosynthesis (IC50s of 3 nM and 1.1 μM in intact leukocytes and human whole blood, respectively) inhibitor. MK-886 is also a non-competitive PPARα antagonist and can induce apoptosis.

Specifications

Chemical Name or Material MK-886
CAS 118414-82-7
Color White
Purity Grade Notes Research
Molecular Formula C27H34ClNO2S
Quantity 25 mg
Synonym L 663536
Solubility Information DMSO : 75 mg/mL (158.87 mM; Need ultrasonic)
SMILES CC(C)C1=CC=C(N(CC2=CC=C(Cl)C=C2)C(CC(C)(C(O)=O)C)=C3SC(C)(C)C)C3=C1
Molecular Weight (g/mol) 472.08
Formula Weight 472.08
Percent Purity 99.11%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.