missing translation for 'onlineSavingsMsg'
Learn More

MK-8033 hydrochloride, MedChemExpress

Product Code. 16405638
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16405638

Brand: Medchem Express HY13299A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MK-8033 hydrochloride is an orally active ATP competitive c-Met/Ron dual inhibitor (IC50s: 1 nM (c-Met),7 nM (Ron)), with preferential binding to the activated kinase conformation. MK-8033 hydrochloride can be used in the research of cancers, such as breast and bladder cancers, non-small cell lung cancers (NSCLCs).

MK-8033 hydrochloride is an orally active ATP competitive c-Met/Ron dual inhibitor (IC50s: 1 nM (c-Met),7 nM (Ron)), with preferential binding to the activated kinase conformation. MK-8033 hydrochloride can be used in the research of cancers, such as breast and bladder cancers, non-small cell lung cancers (NSCLCs).

Specifications

Chemical Name or Material MK-8033 hydrochloride
CAS 1283000-43-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C25H22ClN5O3S
Quantity 10 mg
Solubility Information H2O : 7.14 mg/mL (14.06 mM; Need ultrasonic) ∣DMSO : 5.88 mg/mL (11.58 mM; Need ultrasonic)
SMILES O=S(CC1=CC=C2C(C(C3=CC(C4=CN(C)N=C4)=CN=C3C=C2)=O)=C1)(NCC5=NC=CC=C5)=O.Cl
Molecular Weight (g/mol) 507.99
Formula Weight 507.99
Percent Purity 99.7%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.