missing translation for 'onlineSavingsMsg'
Learn More

MK-8033 hydrochloride, MedChemExpress

Product Code. 16481797
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16481797 50 mg 50mg
16458327 5 mg 5mg
16405638 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16481797 Supplier Medchem Express Supplier No. HY13299A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MK-8033 hydrochloride is an orally active ATP competitive c-Met/Ron dual inhibitor (IC50s: 1 nM (c-Met),7 nM (Ron)), with preferential binding to the activated kinase conformation. MK-8033 hydrochloride can be used in the research of cancers, such as breast and bladder cancers, non-small cell lung cancers (NSCLCs).

MK-8033 hydrochloride is an orally active ATP competitive c-Met/Ron dual inhibitor (IC50s: 1 nM (c-Met),7 nM (Ron)), with preferential binding to the activated kinase conformation. MK-8033 hydrochloride can be used in the research of cancers, such as breast and bladder cancers, non-small cell lung cancers (NSCLCs).

Specifications

Chemical Name or Material MK-8033 hydrochloride
CAS 1283000-43-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C25H22ClN5O3S
Quantity 50 mg
Solubility Information H2O : 7.14 mg/mL (14.06 mM; Need ultrasonic) ∣DMSO : 5.88 mg/mL (11.58 mM; Need ultrasonic)
SMILES O=S(CC1=CC=C2C(C(C3=CC(C4=CN(C)N=C4)=CN=C3C=C2)=O)=C1)(NCC5=NC=CC=C5)=O.Cl
Molecular Weight (g/mol) 507.99
Formula Weight 507.99
Percent Purity 99.7%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.