missing translation for 'onlineSavingsMsg'
Learn More

(-)-MK 801 Maleate, TRC

Product Code. 30466907
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30466907 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30466907 Supplier Toronto Research Chemicals Supplier No. TRCM42504825MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (5R)-5H-Dibenzo[a,d]cyclohepten-5,10-imine-10,11-dihydro-5-methyl-(Z)-2-butenedioate
CAS 121917-57-5
Molecular Formula C16H15N . C4H4O4
Quantity 25 mg
Synonym (-)-MK 801 Maleate, (-)-MK 801
InChI Formula InChI=1S/C16H15N.C4H4O4/c1-16-13-8-4-2-6-11(13)10-15(17-16)12-7-3-5-9-14(12)16;5-3(6)1-2-4(7)8/h2-9,15,17H,10H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1-/t15-,16+;/m0./s1
SMILES C[C@@]12N[C@@H](Cc3ccccc13)c4ccccc24.OC(=O)\C=C/C(=O)O
Molecular Weight (g/mol) 337.4
Formula Weight 337.1314
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.