missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ MK 571
Description
Potent CysLT1 (LTD4) leukotriene receptor inverse agonist (EC50 = 1.3 nM). Antagonizes LTD4-induced contractions of guinea pig trachea and ileum (pA2 values are 9.4 and 10.5 respectively). Also inhibitor of multidrug resistance protein-1 (MRP1) mediated transport; in vitro augments the effects of cytotoxic agents on malignant cells.
Specifications
Specifications
| CAS | 115104-28-4 |
| Quantity | 10 mg |
| Molecular Formula | C26H27ClN2O3S2 |
| Formula Weight | 515.09 |
| Purity | >98% |
| Molecular Weight (g/mol) | 515.083 |
| Chemical Name or Material | MK 571 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?