missing translation for 'onlineSavingsMsg'
Learn More

MK-5108, MedChemExpress

Product Code. 16465588
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16465588 10 mg 10mg
16438287 5 mg 5mg
16431757 50 mg 50mg
16406136 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16465588 Supplier Medchem Express Supplier No. HY13252
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress MK-5108 is a highly potent and specific inhibitor of Aurora A kinase with an IC50 value of 0.064 nM.

MK-5108 is a highly potent and specific inhibitor of Aurora A kinase with an IC50 value of 0.064 nM.

Specifications

Chemical Name or Material MK-5108
CAS 1010085-13-8
Color White
Purity Grade Notes Research
Molecular Formula C22H21ClFN3O3S
Quantity 10 mg
Synonym VX-689
Solubility Information DMSO : 12.5 mg/mL (27.06 mM; Need ultrasonic)
SMILES FC(C(Cl)=CC=C1)=C1O[C@H](CC2)CC[C@@]2(C(O)=O)CC3=CC=CC(NC4=NC=CS4)=N3
Molecular Weight (g/mol) 461.94
Formula Weight 461.94
Percent Purity 96.69%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.