missing translation for 'onlineSavingsMsg'
Learn More

MK-4256, MedChemExpress

Product Code. p-200100104
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16481349 1 mg 1mg
16448397 5 mg 5mg
16436196 100 mg 100mg
16431867 50 mg 50mg
16415708 10 mg 10mg
16413366 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16481349 Supplier Medchem Express Supplier No. HY13466

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MK-4256 is a potent and selective SSTR3 antagonist with IC50s of 0.66 nM and 0.36 nM in human and mouse receptor binding assays, respectively.

MK-4256 is a potent and selective SSTR3 antagonist with IC50s of 0.66 nM and 0.36 nM in human and mouse receptor binding assays, respectively.

Specifications

Chemical Name or Material MK-4256
CAS 1104599-69-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C27H23FN8O
Quantity 1 mg
Solubility Information DMSO : ≥ 100 mg/mL (202.22 mM)
SMILES CN1N=CC([C@@]2(C3=NOC(C)=N3)N[C@@H](C4=NC=C(C5=CC=C(F)C=C5)N4)CC6=C2NC7=C6C=CC=C7)=C1
Molecular Weight (g/mol) 494.52
Formula Weight 494.52
Percent Purity 99.48%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.