missing translation for 'onlineSavingsMsg'
Learn More

MK-4256, MedChemExpress

Product Code. 16413366
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16413366

Brand: Medchem Express HY13466

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MK-4256 is a potent and selective SSTR3 antagonist with IC50s of 0.66 nM and 0.36 nM in human and mouse receptor binding assays, respectively.

MK-4256 is a potent and selective SSTR3 antagonist with IC50s of 0.66 nM and 0.36 nM in human and mouse receptor binding assays, respectively.

Specifications

Chemical Name or Material MK-4256
CAS 1104599-69-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C27H23FN8O
Quantity 25 mg
Solubility Information DMSO : ≥ 100 mg/mL (202.22 mM)
SMILES CN1N=CC([C@@]2(C3=NOC(C)=N3)N[C@@H](C4=NC=C(C5=CC=C(F)C=C5)N4)CC6=C2NC7=C6C=CC=C7)=C1
Molecular Weight (g/mol) 494.52
Formula Weight 494.52
Percent Purity 99.48%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.